siRNA Details
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022181.1 | NP_071517.1 | 4 | 1241 | ||
GenBank mRNA | AF083879.1 | - | 4 | 1241 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022181.1 | NP_071517.1 | 4 | 1241 | ||
GenBank mRNA | AF083879.1 | - | 4 | 1241 |