siRNA Details
| Gene Symbol: Cask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022184.1 | NP_071520.1 | 2 | 497 | ||
| GenBank mRNA | U47110.1 | - | 2 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cask| Gene Symbol: Cask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022184.1 | NP_071520.1 | 2 | 497 | ||
| GenBank mRNA | U47110.1 | - | 2 | 497 | ||