siRNA Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022186.1 | NP_071522.1 | 2 | 214 | ||
| GenBank mRNA | AB024930.1 | - | 2 | 214 | ||
| Gene Symbol: LOC100363472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728074.4 | XP_002728120.1 | 1 | 162 | ||
| Gene Symbol: RGD1560263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574989.5 | XP_574989.1 | 1 | 109 | ||

Nrbf2
Genomic Map