siRNA Details
| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022188.1 | NP_071524.1 | 5 | 680 | ||
| XM_008768533.1 | XP_008766755.1 | 4 | 1570 | |||
| XM_008768534.1 | XP_008766756.1 | 4 | 1570 | |||
| XM_008768535.1 | XP_008766757.1 | 4 | 1570 | |||
| XM_008768536.1 | XP_008766758.1 | 4 | 1570 | |||
| GenBank mRNA | AF041082.1 | - | 5 | 680 | ||

Robo1
Genomic Map