siRNA Details
| Gene Symbol: Htr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022189.1 | NP_071525.1 | 3, 4 | 270 | ||
| GenBank mRNA | AF155044.1 | - | 3, 4 | 270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr3b| Gene Symbol: Htr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022189.1 | NP_071525.1 | 3, 4 | 270 | ||
| GenBank mRNA | AF155044.1 | - | 3, 4 | 270 | ||