siRNA Details
Gene Symbol: Acan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022190.1 | NP_071526.1 | 3, 4 | 837 | ||
GenBank mRNA | J03485.1 | - | 3, 4 | 837 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022190.1 | NP_071526.1 | 3, 4 | 837 | ||
GenBank mRNA | J03485.1 | - | 3, 4 | 837 |