siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022507.1 | NP_071952.1 | 3 | 415 | ||
| XM_008764337.1 | XP_008762559.1 | 3 | 316 | |||
| GenBank mRNA | J04532.1 | - | 3 | 415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcz| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022507.1 | NP_071952.1 | 3 | 415 | ||
| XM_008764337.1 | XP_008762559.1 | 3 | 316 | |||
| GenBank mRNA | J04532.1 | - | 3 | 415 | ||