siRNA Details
| Gene Symbol: Insl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022583.1 | NP_072105.1 | 1 | 265 | ||
| GenBank mRNA | AF159506.1 | - | 1 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insl6| Gene Symbol: Insl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022583.1 | NP_072105.1 | 1 | 265 | ||
| GenBank mRNA | AF159506.1 | - | 1 | 265 | ||