siRNA Details
| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072865.1 | - | 2 | 152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txnrd2| Gene Symbol: Txnrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072865.1 | - | 2 | 152 | ||