siRNA Details
| Gene Symbol: Rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022672.1 | NP_073163.1 | 3 | 199 | ||
| GenBank mRNA | X15040.1 | - | 3 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps14| Gene Symbol: Rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022672.1 | NP_073163.1 | 3 | 199 | ||
| GenBank mRNA | X15040.1 | - | 3 | 199 | ||