siRNA Details
| Gene Symbol: Mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022673.2 | NP_073164.2 | 3 | 532 | ||
| XM_006229566.2 | XP_006229628.1 | 3 | 706 | |||
| XM_006229567.2 | XP_006229629.1 | 2 | 816 | |||
| GenBank mRNA | DQ897368.1 | - | 2 | 408 | ||
| DQ897369.1 | - | 3 | 532 | |||
| M94064.1 | - | 2 | 374 | |||

Mecp2
Genomic Map