siRNA Details
| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022676.3 | NP_073167.1 | 6, 7 | 648 | ||
| XM_006242437.2 | XP_006242499.1 | 6, 7 | 660 | |||
| GenBank mRNA | AJ276593.1 | - | 6, 7 | 521 | ||
| AY648296.1 | - | 6, 7 | 648 | |||
| BC078820.1 | - | 6, 7 | 649 | 7127505 | ||
| FQ217772.1 | - | 6, 7 | 617 | |||
| J05592.1 | - | 6, 7 | 520 | |||

Ppp1r1a
Genomic Map