siRNA Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022710.1 | NP_073201.1 | 3, 4 | 306 | ||
| XM_006242386.2 | XP_006242448.1 | 2, 3 | 878 | |||
| GenBank mRNA | AF327905.1 | - | 3, 4 | 262 | ||
| AF327906.1 | - | 3, 4 | 299 | |||
| FQ212460.1 | - | 3, 4 | 362 | |||
| M94537.1 | - | 3, 4 | 306 | |||

Pde1b
Genomic Map