siRNA Details
| Gene Symbol: Dpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022850.1 | NP_074041.1 | 7, 8 | 943 | ||
| XM_006235880.2 | XP_006235942.1 | 7, 8 | 5016 | |||
| GenBank mRNA | M76426.1 | - | 7, 8 | 943 | ||
| M76427.1 | - | 7, 8 | 796 | |||

Dpp6
Genomic Map