siRNA Details
| Gene Symbol: Pdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022852.3 | NP_074043.3 | 2 | 1275 | ||
| GenBank mRNA | U04833.1 | - | 2 | 1275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdx1| Gene Symbol: Pdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022852.3 | NP_074043.3 | 2 | 1275 | ||
| GenBank mRNA | U04833.1 | - | 2 | 1275 | ||