siRNA Details
| Gene Symbol: Rab7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023950.3 | NP_076440.1 | 6 | 1095 | ||
| XM_006236849.2 | XP_006236911.1 | 6 | 1046 | |||
| GenBank mRNA | BC072470.1 | - | 6 | 1095 | 7098657 | |
| X12535.1 | - | 6 | 967 | |||

Rab7a
Genomic Map