siRNA Details
| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024130.1 | NP_077044.1 | 2 | 422 | ||
| GenBank mRNA | FQ223809.1 | - | 178 | |||
| X62160.1 | - | 2 | 422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dctn1| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024130.1 | NP_077044.1 | 2 | 422 | ||
| GenBank mRNA | FQ223809.1 | - | 178 | |||
| X62160.1 | - | 2 | 422 | |||