siRNA Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109986.1 | NP_001103456.1 | 4 | 459 | ||
NM_024134.2 | NP_077048.2 | 3 | 411 | |||
XM_006241444.2 | XP_006241506.1 | 5 | 1637 | |||
XM_006241445.2 | XP_006241507.1 | 4 | 1589 | |||
GenBank mRNA | AW916370.1 | - | 4 | 459 | ||
BC100664.1 | - | 4 | 422 | 7932840 | ||
CF977695.1 | - | 3 | 414 | |||
U30186.1 | - | 3 | 403 | |||
U36994.1 | - | 3 | 381 |
