siRNA Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109986.1 | NP_001103456.1 | 4 | 469 | ||
| NM_024134.2 | NP_077048.2 | 3 | 421 | |||
| XM_006241444.2 | XP_006241506.1 | 5 | 1647 | |||
| XM_006241445.2 | XP_006241507.1 | 4 | 1599 | |||
| GenBank mRNA | AW916370.1 | - | 4 | 469 | ||
| BC100664.1 | - | 4 | 432 | 7932840 | ||
| CF977695.1 | - | 3 | 424 | |||
| U30186.1 | - | 3 | 413 | |||
| U36994.1 | - | 3 | 391 | |||

Ddit3
Genomic Map