siRNA Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024135.2 | NP_077049.2 | 16 | 3093 | ||
| XM_006251236.2 | XP_006251298.1 | 15 | 4410 | |||
| GenBank mRNA | BC065578.1 | - | 15 | 3278 | 5623751 | |
| D31874.1 | - | 16 | 3093 | |||
| D31875.1 | - | 15 | 3322 | |||
| D31876.1 | - | 16 | 3341 | |||
| D31877.1 | - | 16 | 3465 | |||

Limk2
Genomic Map