siRNA Details
| Gene Symbol: Gng7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024138.1 | NP_077052.1 | 3 | 575 | ||
| GenBank mRNA | DQ120497.1 | - | 2 | 333 | ||
| DQ120498.1 | - | 2 | 333 | |||
| FQ211653.1 | - | 555 | ||||
| L23219.1 | - | 3 | 575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gng7| Gene Symbol: Gng7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024138.1 | NP_077052.1 | 3 | 575 | ||
| GenBank mRNA | DQ120497.1 | - | 2 | 333 | ||
| DQ120498.1 | - | 2 | 333 | |||
| FQ211653.1 | - | 555 | ||||
| L23219.1 | - | 3 | 575 | |||