siRNA Details
| Gene Symbol: Chrna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024354.1 | NP_077330.1 | 5 | 1310 | ||
| GenBank mRNA | AF007212.1 | - | 5 | 1310 | ||
| L31620.1 | - | 1311 | ||||
| M15682.1 | - | 5 | 1289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna4| Gene Symbol: Chrna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024354.1 | NP_077330.1 | 5 | 1310 | ||
| GenBank mRNA | AF007212.1 | - | 5 | 1310 | ||
| L31620.1 | - | 1311 | ||||
| M15682.1 | - | 5 | 1289 | |||