siRNA Details
| Gene Symbol: Notch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024358.1 | NP_077334.1 | 34 | 7994 | ||
| GenBank mRNA | M93661.1 | - | 34 | 7994 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Notch2| Gene Symbol: Notch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024358.1 | NP_077334.1 | 34 | 7994 | ||
| GenBank mRNA | M93661.1 | - | 34 | 7994 | ||