siRNA Details
| Gene Symbol: IFIT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012420.2 | NP_036552.1 | 2 | 317 | ||
| GenBank mRNA | AK293346.1 | - | 2 | 123 | ||
| AK312358.1 | - | 2 | 150 | |||
| BC025786.1 | - | 2 | 296 | 5229306 | ||
| CR457031.1 | - | 1 | 86 | |||
| DC392439.1 | - | 2 | 317 | |||
| U34605.1 | - | 2 | 203 | |||

IFIT5
Genomic Map