siRNA Details
| Gene Symbol: Hes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024360.3 | NP_077336.3 | 4 | 890 | ||
| GenBank mRNA | BC061730.1 | - | 4 | 890 | 5598665 | |
| D13417.1 | - | 4 | 928 | |||
| L04527.1 | - | 1452 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hes1| Gene Symbol: Hes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024360.3 | NP_077336.3 | 4 | 890 | ||
| GenBank mRNA | BC061730.1 | - | 4 | 890 | 5598665 | |
| D13417.1 | - | 4 | 928 | |||
| L04527.1 | - | 1452 | ||||