siRNA Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030828.1 | NP_110455.1 | 9 | 2215 | ||
| GenBank mRNA | L02896.1 | - | 9 | 2215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc1| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030828.1 | NP_110455.1 | 9 | 2215 | ||
| GenBank mRNA | L02896.1 | - | 9 | 2215 | ||