siRNA Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030829.1 | NP_110456.1 | 2 | 271 | ||
| XM_006231686.1 | XP_006231748.1 | 2 | 324 | |||
| GenBank mRNA | BC081792.1 | - | 2 | 324 | 7123515 | |
| U34841.1 | - | 2 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grk5| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030829.1 | NP_110456.1 | 2 | 271 | ||
| XM_006231686.1 | XP_006231748.1 | 2 | 324 | |||
| GenBank mRNA | BC081792.1 | - | 2 | 324 | 7123515 | |
| U34841.1 | - | 2 | 271 | |||