siRNA Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030829.1 | NP_110456.1 | 14, 15 | 1730 | ||
| XM_006231686.1 | XP_006231748.1 | 14, 15 | 1783 | |||
| GenBank mRNA | BC081792.1 | - | 14, 15 | 1783 | 7123515 | |
| U34841.1 | - | 14, 15 | 1730 | |||

Grk5
Genomic Map