siRNA Details
| Gene Symbol: Sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030996.1 | NP_112258.1 | 4 | 693 | ||
| GenBank mRNA | AF004218.1 | - | 4 | 693 | ||
| AF067769.1 | - | 4 | 745 | |||
| AF087827.1 | - | 4 | 698 | |||
| BC061978.1 | - | 4 | 682 | 5623693 | ||

Sigmar1
Genomic Map