siRNA Details
| Gene Symbol: Prl3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031316.1 | NP_112606.1 | 4 | 443 | ||
| GenBank mRNA | AB019119.1 | - | 4 | 443 | ||
| AB019945.1 | - | 4 | 434 | |||
| AF150741.1 | - | 4 | 443 | |||
| AF234638.1 | - | 4 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl3c1| Gene Symbol: Prl3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031316.1 | NP_112606.1 | 4 | 443 | ||
| GenBank mRNA | AB019119.1 | - | 4 | 443 | ||
| AB019945.1 | - | 4 | 434 | |||
| AF150741.1 | - | 4 | 443 | |||
| AF234638.1 | - | 4 | 434 | |||