siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031558.3 | NP_113746.1 | 3 | 340 | ||
| GenBank mRNA | AB001349.1 | - | 296 | |||
| AB006007.1 | - | 3 | 341 | |||
| AF044081.1 | - | 280 | ||||
| AY736357.1 | - | 3 | 221 | |||
| BC088859.1 | - | 3 | 232 | 7311249 | ||
| EF091859.1 | - | 2 | 87 | |||

Star
Genomic Map