siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB001349.1 | - | 1191 | |||
| AF044081.1 | - | 1175 | ||||
| BC088859.1 | - | 7 | 1127 | 7311249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Star| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB001349.1 | - | 1191 | |||
| AF044081.1 | - | 1175 | ||||
| BC088859.1 | - | 7 | 1127 | 7311249 | ||