siRNA Details
| Gene Symbol: Cpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031559.2 | NP_113747.2 | 2 | 131 | ||
| XM_006230695.1 | XP_006230757.1 | 3 | 241 | |||
| GenBank mRNA | BC072522.1 | - | 2 | 131 | 7105825 | |
| L07736.1 | - | 2 | 185 | |||
| U88294.1 | - | 1 | 83 | |||

Cpt1a
Genomic Map