siRNA Details
| Gene Symbol: Ctsk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031560.2 | NP_113748.1 | 6 | 836 | ||
| GenBank mRNA | AF010306.1 | - | 6 | 805 | ||
| BC078793.1 | - | 6 | 836 | 7134253 | ||
| FQ222438.1 | - | 6 | 780 | |||
| FQ228691.1 | - | 6 | 853 | |||

Ctsk
Genomic Map