siRNA Details
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031569.3 | NP_113757.1 | 2 | 345 | ||
XM_006235736.2 | XP_006235798.1 | 3 | 435 | |||
XM_006235738.2 | XP_006235800.1 | 2 | 193 | |||
XM_006235739.2 | XP_006235801.1 | 3 | 435 | |||
GenBank mRNA | AF115267.1 | - | 1 | 132 | ||
AY152731.1 | - | 1 | 104 | |||
D16438.1 | - | 2 | 156 | |||
L28144.1 | - | 2 | 276 | |||
L29419.1 | - | 1 | 239 | |||
L33916.1 | - | 2 | 210 | |||
U01913.1 | - | 2 | 285 | |||
U05239.1 | - | 2 | 287 | |||
U07871.1 | - | 1 | 135 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
