siRNA Details
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031584.2 | NP_113772.1 | 2 | 546 | ||
GenBank mRNA | D83044.1 | - | 2 | 549 | ||
X98334.1 | - | 2 | 495 | |||
Y13154.1 | - | 2 | 496 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031584.2 | NP_113772.1 | 2 | 546 | ||
GenBank mRNA | D83044.1 | - | 2 | 549 | ||
X98334.1 | - | 2 | 495 | |||
Y13154.1 | - | 2 | 496 |