siRNA Details
| Gene Symbol: HACL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284415.1 | NP_001271344.1 | 4 | 629 | ||
| NM_001284416.1 | NP_001271345.1 | 4 | 629 | |||
| NM_012260.3 | NP_036392.2 | 4 | 629 | |||
| NR_104315.1 | 4 | 629 | ||||
| GenBank mRNA | AF161397.1 | - | 4 | 249 | ||
| AJ131753.1 | - | 4 | 339 | |||
| AK301990.1 | - | 4 | 364 | |||
| AK302086.1 | - | 4 | 346 | |||
| BC001627.1 | - | 4 | 347 | 3538159 | ||
| BC007440.2 | - | 4 | 334 | |||
| DA609383.1 | - | 4 | 346 | |||
| DB049608.1 | - | 4 | 400 | |||

HACL1
Genomic Map