siRNA Details
| Gene Symbol: Sart1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031596.1 | NP_113784.1 | 1 | 348 | ||
| GenBank mRNA | AB014722.1 | - | 1 | 348 | ||
| BC083551.1 | - | 1 | 321 | 7134496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sart1| Gene Symbol: Sart1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031596.1 | NP_113784.1 | 1 | 348 | ||
| GenBank mRNA | AB014722.1 | - | 1 | 348 | ||
| BC083551.1 | - | 1 | 321 | 7134496 | ||