siRNA Details
| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031597.4 | NP_113785.3 | 1 | 718 | ||
| GenBank mRNA | AF091247.1 | - | 2 | 264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq3| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031597.4 | NP_113785.3 | 1 | 718 | ||
| GenBank mRNA | AF091247.1 | - | 2 | 264 | ||