siRNA Details
| Gene Symbol: Kcnj10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031602.1 | NP_113790.1 | 2 | 357 | ||
| XM_006250285.2 | XP_006250347.1 | 2 | 1646 | |||
| XM_008769737.1 | XP_008767959.1 | 2 | 1086 | |||
| GenBank mRNA | U27558.1 | - | 2 | 357 | ||
| X83585.1 | - | 1 | 237 | |||
| X86818.1 | - | 2 | 355 | |||

Kcnj10
Genomic Map