siRNA Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031604.2 | NP_113792.2 | 4 | 441 | ||
| XM_006247295.2 | XP_006247357.1 | 4 | 441 | |||
| XM_006247296.2 | XP_006247358.1 | 4 | 441 | |||
| XM_006247297.2 | XP_006247359.1 | 4 | 441 | |||
| XM_008768002.1 | XP_008766224.1 | 4 | 404 | |||
| XM_008768003.1 | XP_008766225.1 | 4 | 404 | |||
| XM_008768004.1 | XP_008766226.1 | 4 | 442 | |||
| GenBank mRNA | DQ286421.1 | - | 3 | 283 | ||
| DQ286422.1 | - | 3 | 283 | |||
| DQ286423.1 | - | 3 | 283 | |||
| DQ286424.1 | - | 3 | 283 | |||
| M58758.1 | - | 3 | 313 | |||

Atp6v0a1
Genomic Map