siRNA Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031604.2 | NP_113792.2 | 22 | 3619 | ||
| XM_006247295.2 | XP_006247357.1 | 22 | 3640 | |||
| XM_006247296.2 | XP_006247358.1 | 21 | 3622 | |||
| XM_006247297.2 | XP_006247359.1 | 21 | 3601 | |||
| XM_008768002.1 | XP_008766224.1 | 22 | 3603 | |||
| XM_008768003.1 | XP_008766225.1 | 22 | 3582 | |||
| GenBank mRNA | CB586356.1 | - | 1 | 511 | ||
| M58758.1 | - | 21 | 3489 | |||

Atp6v0a1
Genomic Map