siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031606.1 | NP_113794.1 | 5 | 315 | ||
| GenBank mRNA | AF017185.1 | - | 5 | 315 | ||
| AF455569.1 | - | 5 | 315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pten| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031606.1 | NP_113794.1 | 5 | 315 | ||
| GenBank mRNA | AF017185.1 | - | 5 | 315 | ||
| AF455569.1 | - | 5 | 315 | |||