siRNA Details
| Gene Symbol: Gria1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031608.1 | NP_113796.1 | 2 | 405 | ||
| GenBank mRNA | M36418.2 | - | 2 | 208 | ||
| X17184.1 | - | 2 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gria1| Gene Symbol: Gria1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031608.1 | NP_113796.1 | 2 | 405 | ||
| GenBank mRNA | M36418.2 | - | 2 | 208 | ||
| X17184.1 | - | 2 | 405 | |||