siRNA Details
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031608.1 | NP_113796.1 | 7 | 1306 | ||
GenBank mRNA | M36418.2 | - | 7 | 1109 | ||
M38060.1 | - | 7 | 1109 | |||
X17184.1 | - | 7 | 1306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031608.1 | NP_113796.1 | 7 | 1306 | ||
GenBank mRNA | M36418.2 | - | 7 | 1109 | ||
M38060.1 | - | 7 | 1109 | |||
X17184.1 | - | 7 | 1306 |