siRNA Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031610.3 | NP_113798.1 | 1 | 171 | ||
| GenBank mRNA | L25264.1 | - | 1 | 171 | ||
| U01071.1 | - | 1 | 144 | |||
| U01141.1 | - | 1 | 185 | |||
| U09243.1 | - | 299 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj3| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031610.3 | NP_113798.1 | 1 | 171 | ||
| GenBank mRNA | L25264.1 | - | 1 | 171 | ||
| U01071.1 | - | 1 | 144 | |||
| U01141.1 | - | 1 | 185 | |||
| U09243.1 | - | 299 | ||||