siRNA Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031614.2 | NP_113802.2 | 4 | 339 | ||
| GenBank mRNA | AF108213.1 | - | 3 | 181 | ||
| AF220760.1 | - | 3 | 235 | |||
| AF220761.1 | - | 3 | 213 | |||
| BC085726.1 | - | 436 | 7112649 | |||
| U63923.2 | - | 3 | 230 | |||

Txnrd1
Genomic Map