siRNA Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031614.2 | NP_113802.2 | 8, 9 | 1006 | ||
| GenBank mRNA | AF108213.1 | - | 7, 8 | 848 | ||
| AF220760.1 | - | 7, 8 | 902 | |||
| BC085726.1 | - | 1103 | 7112649 | |||
| FQ225805.1 | - | 5, 6 | 651 | |||
| U63923.2 | - | 7, 8 | 897 | |||

Txnrd1
Genomic Map