siRNA Details
| Gene Symbol: Csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031617.1 | NP_113805.1 | 4 | 349 | ||
| XM_006242038.2 | XP_006242100.1 | 5 | 498 | |||
| GenBank mRNA | AB042191.1 | - | 4 | 349 | ||
| AB042192.1 | - | 4 | 349 | |||
| AB056113.1 | - | 4 | 349 | |||

Csnk1e
Genomic Map