siRNA Details
| Gene Symbol: Phgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031620.1 | NP_113808.1 | 6 | 549 | ||
| XM_006233010.1 | XP_006233072.1 | 5 | 572 | |||
| GenBank mRNA | BC086327.1 | - | 5 | 577 | 7115765 | |
| X97772.1 | - | 5 | 508 | |||

Phgdh
Genomic Map